missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD47 Antibody (letaplimab) - Humanized, IgG4SP, Novus Biologicals™

Human Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-27991-100ug

 View more versions of this product

Product Code. 30046382

  • £247.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD47
Monoclonal
Unconjugated
Q08722
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG4
Flow Cytometry, ELISA, Functional Assay
letaplimab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
CD47
100 μg
Immunology, Signal Transduction
961
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.