Learn More
CD46 Monoclonal Antibody (M177), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA140183
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-40183 detects CD46 from human samples. MA1-40183 has been successfully used in flow cytometry, immunoprecipitation, functional assays, immunohistochemistry (frozen) and Western blot applications. The MA1-40183 immunogen is purified MCP. Endotoxin <24 EU/mg. Positive control: CD46 is expressed on every cell and tissue; Negative control: Erythrocytes.
CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.Specifications
CD46 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and no preservative | |
P15529 | |
CD46 | |
purified MCP | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
M177 | |
Unconjugated | |
CD46 | |
AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antig | |
Mouse | |
Protein G | |
RUO | |
4179 | |
4° C | |
Liquid |