Learn More
Invitrogen™ CD46 Monoclonal Antibody (M037)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547651
Description
Clone M037 mouse monoclonal antibody detects a 45-60 kDa* protein on SDS-PAGE Native immunoblots of human A549, MeWo, MDA-MB-231, and MCF7 cells, as well as human lung tissues. This antibody weakly detects denatured CD46. The antibody works for western blot, immunoprecipitation, ELISA capture, and immunocytochemistry.
CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.
Specifications
| CD46 | |
| Monoclonal | |
| 0.25 mg/mL | |
| PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P15529 | |
| CD46 | |
| Clone (M037) was generated from a proprietary antigen related to native human CD46 expressed in NCI-H1915 lung cancer cell line. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M037 | |
| Unconjugated | |
| CD46 | |
| AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antigen | |
| Mouse | |
| Protein G | |
| RUO | |
| 4179 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.