missing translation for 'onlineSavingsMsg'
Learn More

CD46 Antibody (MM.1), Novus Biologicals™

Product Code. 18382863 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382863 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382863 Supplier Novus Biologicals Supplier No. NBP311191

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD46 Monoclonal specifically detects CD46 in Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD46
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone MM.1
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
Host Species Mouse
Immunogen Second and third SCR domains of MCP fused to IgG-Fc; MCP23-Ig
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4179
Target Species Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.