missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RO Mouse, Clone: UCHL-1, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 5788-MSM1-P1
This item is not returnable.
View return policy
Specifications
| CD45RO | |
| Cell surface | |
| UCHL-1 | |
| Unconjugated | |
| NP_002829 | |
| Mouse | |
| 180-185kDa | |
| Protein A/G Purified | |
| RUO | |
| 5788 | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| PTPRC | |
| Cultured human T-cells from an IL-2-dependent T-cell line (CA1) | |
| CCRF-CEM, Jurkat or MOLT-4 cells. Paracortex in a tonsil or a reactive lymph node. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction