missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD45RO Fab Streptamer™ Isolation Kit MB
CD45RO Fab Streptamer Isolation Kit MB is for reversible cell isolation delivering label-free target cells.
Brand: IBA Lifesciences 6-8000-209
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Strep-Tactin™ Magnetic Microbeads for Fab Streptamers™; 2 vials for 5 x 108 cells each
- CD45RO Fab-Strep, human for isolation of T-cells; 5 vials for 2 x 108 cells
- Biotin stock solution for Streptamer™ technology; 2 x 1mL
- Buffer IS, Streptamer™ Washing Buffer, 10x concentrated; 50mL
- Microbeads are intended for use with a permanent magnet for manual cell isolation; we recommend our StrepMan magnet for 2 x 15mL and 2 x 50mL Falcon tubes.
- Principle of Selection: Isolation of human CD45RO T cells is performed by positive selection.
- PBMCs are labeled with magnetic CD45RO Fab Streptamers™.
- Labeled cells are isolated in a strong magnet (StrepMan, horizontal position) where they migrate toward the tube wall on the side of the magnet.
- This CD45RO positive cell fraction is collected and cells are liberated from all labeling reagents by addition of biotin in a strong magnet (StrepMan, upright position).
- The magnetic Streptamers™ migrate toward the tube wall and the label-free cells remain in the supernatant.
- Biotin is removed by washing.
- The resulting cell preparation is highly enriched with CD45RO+ T cells with a purity of more than 90%.
- No depletion steps and no columns are needed.
Specifications
PBMCs | |
Magnetic Microspheres Coated with Strep-Tactin | |
1 x 109 |