Learn More
CD45RO Antibody (UCHL-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-29631
 View more versions of this product
Product Code. 18736123
  • £439.00 / 0.10mg
Description
CD45RO Monoclonal specifically detects CD45RO in Human, Chimpanzee, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry 0.5-1 μg/mL, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 0.5-1 μg/mL, Immunohistochemistry-Frozen 0.5-1 μg/mL, Dual RNAscope ISH-IHC
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
182 kDa
Protein A or G purified