missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RC Rat anti-Mouse, BV750, Clone: DNL-1.9, BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 747146
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
CD45RC Monoclonal antibody specifically detects CD45RC in Mouse samples. It is validated for Flow Cytometry
Tekniske data
| CD45RC | |
| Monoclonal | |
| Brilliant Violet 750 | |
| Ptprc; Cd45; CD45R; L-CA; Ly-5; Lyt-4; T200 | |
| 70Z/3 mouse pre-B cell line | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| DNL-1.9 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion