Learn More
CD45RC Mouse Anti-Rat, Brilliant Violet 786, Clone: OX-22, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742491
Description
The OX-22 antibody reacts with CD45RC on pre-B lymphocytes, B cells, CD8+ T suppressor/cytotoxic cells, and a subset of CD4+ T helper (Th) lymphocytes. It weakly reacts with thymocytes. CD45RC is a high-molecular-weight isoform of CD45 (Leukocyte Common Antigen); its level of expression distinguishes subpopulations of CD4+ T cells with Th1-like and Th2-like effector functions. Levels of expression of CD45RC have also been reported to distinguish resting from activated T cells at various stages of maturation. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
Specifications
| CD45RC | |
| OX-22 | |
| Brilliant Violet 786 | |
| CD45RC | |
| PHA-activated rat lymph node cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.