missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD45RC Monoclonal Antibody (OX-22)

Mouse Monoclonal Antibody
£239.00 - £414.00
Specifications
| Antigen | CD45RC |
|---|---|
| Clone | OX-22 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11584403
![]() |
Invitrogen™
MA170013 |
250 μg |
£239.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15358360
|
Invitrogen™
MA517457 |
500 μg |
£414.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RC Monoclonal Antibody for IHC (F), Flow
CD45RC (PTPRC) is a protein tyrosine-protein phosphatase required for T-cell activation through the antigen receptor. It acts as a positive regulator of T-cell coactivation upon binding to DPP4. The first PTPase domain has enzymatic activity, the second affects the substrate specificity of the first. Upon T-cell activation, CD45RC recruits and dephosphorylates SKAP1 and FYN. Mutations of the gene can result in severe combined immunodeficiency and multiple sclerosis.Specifications
| CD45RC | |
| 1 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.02% sodium azide | |
| P04157 | |
| 24699 | |
| Phytohaemagglutinin-activated rat lymph node cells. | |
| Antibody |
| OX-22 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Rat | |
| Ptprc | |
| B220; CD45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; LCA; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
| Ptprc | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title