Learn More
CD45RA Mouse anti-Human, Clone: 111-1C5, Abnova™
Mouse monoclonal antibody raised against native human CD45RA.
Brand: Abnova MAB13411.100ug
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains, and thus belongs to receptor type PTP. This gene is specifically expressed in hematopoietic cells. This PTP has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. It functions through either direct interaction with components of the antigen receptor complexes, or by activating various Src family kinases required for the antigen receptor signaling. This PTP also suppresses JAK kinases, and thus functions as a regulator of cytokine receptor signaling. Four alternatively spliced transcripts variants of this gene, which encode distinct isoforms, have been reported. [provided by RefSeq]
Specifications
| CD45RA | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| B220/CD45/CD45R/GP180/LCA/LY5/T200 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 5788 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 111-1C5 | |
| Mouse monoclonal antibody raised against native human CD45RA. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| PTPRC | |
| Stimulated human leukocytes. | |
| 100 μg | |
| Primary | |
| Recognizes a protein of 205 kDa-220 kDa, identified as CD45RA (Workshop III). CD45RA is isoforms of the human leukocyte common antigen (CD45). Human CD45 contains three exons which encode peptide segments designated A, B and C, respectively. The differential splicing of the exons generates at least five isoforms, ABC, AB, BC, B and O. This antibody reacts with ABC and BC isoforms. CD45RA is expressed on 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells, B cells, and leukemic B cell lines. T cells expressing CD45RA are naive or virgin T cells. T cells expressing CD45RO are memory T cells. CD45RA and CD45RO define complementary, predominantly non-overlapping populations of resting peripheral T cells. This monoclonal antibody is useful in study on the subpopulation of CD4+ or CD8+ T cells. It can especially be used to differentiate T cell lymphomas (CD45RO+ve) from B cell lymphomas (CD45RA+ve). | |
| Store at 4°C. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.