missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RA Antibody (PTPRC/4367R), Novus Biologicals™
Rabbit Monoclonal Antibody
£224.00 - £412.00
Specifications
| Antigen | CD45RA |
|---|---|
| Clone | PTPRC/4367R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602134
|
Novus Biologicals
NBP3-20377-20ug |
20 μg |
£224.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614364
|
Novus Biologicals
NBP3-20377-100ug |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CD45RA | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA | |
| 5788 | |
| IgG κ | |
| Protein A or G purified |
| PTPRC/4367R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Recombinant fragment (around aa1-600) of human CD45RA protein (extracellular domain) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title