missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD45RA |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTH-74D4 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide |
| Host Species | Rabbit |
| Immunogen | This antibody was raised by immunising mice with an EBV-transformed B cell line from a patient with MHC class II deficiency. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?