missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45R (B220) Rat, anti-Mouse, Alexa Fluor 488, Clone: RA3-6B2, STEMCELL Technologies
Rat Monoclonal Antibody
£171.45 - £386.10
Specifications
| Antigen | CD45R (B220) |
|---|---|
| Clone | RA3-6B2 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17188501
|
Stemcell Technologies
60019AD.1 |
25 μg |
£171.45
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17178501
|
Stemcell Technologies
60019AD |
100 μg |
£386.10
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The RA3-6B2 antibody reacts with an epitope on the extracellular domain of the exon A‐restricted isoform of mouse CD45
- CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
- The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains (one of which is enzymatically active) that are involved in modulating the function of Src kinases such as Lck and Fyn
- Several isoforms of CD45 have been identified, the expression of which differs according to cell type and functional status
- Alternative splicing of three exons encoding the extracellular RA, RB, and RC polypeptide sequences gives rise to up to 8 isoforms with molecular masses in the range of 200 - 220 kDa
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse B Cell Isolation Kit (Catalog #19854)
Specifications
| CD45R (B220) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 488 | |
| Feline, Human, Mouse | |
| Primary |
| RA3-6B2 | |
| Monoclonal | |
| Rat | |
| IgG2a κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title