missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD45R (B220) Monoclonal Antibody (YKIX753.22.2)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA528396
This item is not returnable.
View return policy
Description
This antibody recognizes canine CD45R and was clustered at the First Canine Leucocyte Antigen Workshop (CLAW). It precipitates an antigen of 220 kDa from Con A Blasts.
CD45R (PTPRC) is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45 contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. CD45 is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists in multiple isoforms as a result of alternative splicing, and these isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. Further, these CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. In addition to its role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. Diseases associated with CD45 dysfunction include Severe Combined Immunodeficiency (T-cell negative, B-Cell Negative) and Hepatitis C.
Specifications
| CD45R (B220) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| 0 | |
| PTPRC | |
| Canine thymocytes. | |
| 100 μg | |
| Primary | |
| Canine | |
| Antibody | |
| IgG2b |
| Cytotoxicity Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| YKIX753.22.2 | |
| Unconjugated | |
| PTPRC | |
| B220; Cd45; CD45 antigen; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen | |
| Rat | |
| Protein G | |
| RUO | |
| 490255 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction