Learn More
CD45 Rat, anti-Mouse, Unconjugated, Clone: 30-F11, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech 65087-1-IG-100UG
Description
CD45, also known as protein tyrosine phosphatase, receptor type C, is a type I transmembrane protein expressed on the surface of all haematopoietic cells with the exception of erythrocytes and platelets (PMID: 3489673; 28615666). CD45 is a pan-haematopoietic cell marker and has been shown to be essential for T- and B-cell activation and signalling (PMID: 9429890; 16378097).
Specifications
| CD45 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| 30-F11 | |
| Unconjugated | |
| 100 μg | |
| 19264 | |
| Liquid |