missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rat anti-Mouse , PE, Clone: I3/2.3 (also known as I3/2 or I3), BD Biosciences™
Rat Monoclonal Antibody
Brand: BD Biosciences 567111
This item is not returnable.
View return policy
Description
CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD45 | |
| Monoclonal | |
| PE | |
| Cd45; L-CA; LCA; Ly-5; Ly5; Ptprc | |
| Mouse Lymphoma Cell Line | |
| RUO | |
| Mouse | |
| IgG2b λ |
| Flow Cytometry | |
| I3/2.3 (also known as I3/2 or I3) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction