missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Rat, anti-Mouse, FITC, Clone: 30-F11, STEMCELL Technologies
Rat Monoclonal Antibody
POA - £94.50
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 30-F11 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17108631
|
Stemcell Technologies
60030FI.1 |
50 μg |
£94.50
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17198621
|
Stemcell Technologies
60030FI |
500 μg | |||||||
Description
- The 30-F11 antibody reacts with an extracellular epitope found on all isoforms and both alloantigens (CD45.1 and CD45.2) of mouse CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets and plasma cells; expression of CD45 is lost during differentiation of these cell types
- CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation and oncogenic transformation
- The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains, one which is enzymatically active, that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
- Several isoforms of CD45 have been identified, the expression of which differs according to cell type and functional status
Specifications
| CD45 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| FITC | |
| Mouse | |
| Primary |
| 30-F11 | |
| Monoclonal | |
| Rat | |
| IgG2b κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title