missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, PE-Cy5, Clone: 2D1, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB97-PE5-25Tests
Additional Details : Weight : 0.01000kg
Specifications
CD45 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
PTPRC | |
Protein A/G Purified | |
RUO | |
5788 | |
IgG1 κ |
Flow Cytometry | |
2D1 | |
PE-Cyanine5 | |
NP_002829 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |