missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, anti-Human, Pacific Blue, Clone: HI30, STEMCELL Technologies
Mouse Monoclonal Antibody
£225.45 - £506.25
Specifications
| Antigen | CD45 |
|---|---|
| Clone | HI30 |
| Applications | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Pacific Blue |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17108501
|
Stemcell Technologies
60018PB.1 |
25 μg |
£225.45
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17198491
|
Stemcell Technologies
60018PB |
100 μg |
£472.50
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17118501
|
Stemcell Technologies
60018PB.2 |
100 Tests |
£506.25
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The HI30 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell types
- CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation
- The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052) and EasySep™ Human CD45 Depletion Kit II (Catalog #17898; partial blocking may be observed)
Specifications
| CD45 | |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| Pacific Blue | |
| Chimpanzee, Human | |
| Primary |
| HI30 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title