missing translation for 'onlineSavingsMsg'
Learn More

CD45 / LCA Mouse, Clone: F10-89-4, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 5788-MSM22-P1

 View more versions of this product

Product Code. 15920758

  • £423.29 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

CD45 / LCA
Cell surface and cytoplasmic
F10-89-4
Unconjugated
NP_002829
Mouse
180-220kDa
Protein A/G Purified
RUO
5788
IgG2a κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
PTPRC
Human T-Lymphocytes
Ramos, U-698, or GA-10 cells. Tonsil
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.