missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 / LCA Mouse, Clone: F10-89-4, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 5788-MSM22-P0
This item is not returnable.
View return policy
Specifications
| CD45 / LCA | |
| Cell surface and cytoplasmic | |
| F10-89-4 | |
| Unconjugated | |
| NP_002829 | |
| Mouse | |
| 180-220kDa | |
| Protein A/G Purified | |
| RUO | |
| 5788 | |
| IgG2a κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| PTPRC | |
| Human T-Lymphocytes | |
| Ramos, U-698, or GA-10 cells. Tonsil | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction