missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Antibody (YKIX716.13), FITC, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-65906F
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Canine, Equine samples. It is validated for Flow Cytometry.
Specifications
| CD45 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PTPRC | |
| Canine thymocytes | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| Recognizes canine CD45 also known as leukocyte common antigen lustered as Canine CD45 in the First Canine Leukocyte Antigen Workshop. CD45 is expressed on all leukocytes in canine peripheral blood and may be against CD45RB isoform. This antibody reacts with CD45 on all outbred mongrels and beagles tested. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| YKIX716.13 | |
| Flow Cytometry | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Canine, Equine | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction