missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD45 Monoclonal specifically detects CD45 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular), CyTOF-ready.
Specifications
Specifications
| Classification | Monoclonal |
| Target Species | Human |
| Content And Storage | Store at 4C. Do not freeze. |
| Form | Purified |
| Isotype | IgG1 κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?