missing translation for 'onlineSavingsMsg'
Learn More

CD45 Antibody (EM-05), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18424633 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424633 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424633 Supplier Novus Biologicals Supplier No. NBP144763AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Flow (Cell Surface), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow Cytometry (Cell Surface)
Classification Monoclonal
Clone EM-05
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen
Host Species Rat
Immunogen This CD45 Antibody (EM-05) was prepared from Murine peripheral blood leukocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.