Learn More
CD45 Antibody (2B11 + PD7/26) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-44857
Product Code. 18630336
  • £391.00 / 7mL
Description
CD45 Monoclonal antibody specifically detects CD45 in Human, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD45
Monoclonal
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human, Canine
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
2B11 + PD7/26
Immunohistochemistry, Immunohistochemistry-Paraffin 0.5 - 1.0 μg/mL
B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen
Isolated neoplastic cells from T cell lymphoma (2B11), and human peripheral blood lymphocytes maintained in T cell growth factor (PD7/26)
7 mL
Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
5788
Store at 4°C.
IgG1 κ