Learn More
CD45.2 Mouse, anti-Mouse, Biotin, Clone: 104, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60118BT
This item is not returnable.
View return policy
Description
- The 104 monoclonal antibody reacts with mouse alloantigen CD45.2 (Ly5.2) expressed by all hematopoietic cells except mature erythrocytes and platelets in mouse strains A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C57BR, C57L, C58, DBA/1, DBA/2, NZB, SWR, and 129
- Clone 104 does not react with leukocytes that express CD45.1 alloantigen
- CD45.2 is an allelic form of CD45 (Leukocyte common antigen), a 180 - 240 kD type I transmembrane glycoprotein
- CD45 is a member of the protein tyrosine phosphatase (PTP) family and functions in a number of immunoregulatory processes, including cell activation, growth, lymphocyte development, differentiation, and oncogenic transformation
- The isoforms of CD45 correlate to specific cell types, as well specific activation and maturation states of immune cells
- The 104 monoclonal antibody has been used extensively in adoptive cell transfer and bone marrow transplantation studies in mice, allowing for the differentiation of CD45.2- and CD45.1- expressing cells
Specifications
| CD45.2 | |
| Monoclonal | |
| Biotin | |
| 500 μg | |
| Mouse | |
| Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Lymphocytes, Myeloid Cells |
| Flow Cytometry | |
| 104 | |
| Mouse | |
| Primary | |
| IgG2a κ |