missing translation for 'onlineSavingsMsg'
Learn More

CD45.1 Mouse, APC-Cy7, Clone: A20, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB42-APC7-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952008

  • £219.00 / 100µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CD45.1
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2a κ
Flow Cytometry
A20
APC-Cyanine7
NP_001255215
Mouse
100 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers