missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45.1 Mouse anti-Mouse, APC-R700, Clone: A20 , BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 565813
This item is not returnable.
View return policy
Description
CD45.1 Monoclonal antibody specifically detects CD45.1 in Mouse samples. It is validated for Flow cytometry.
Specifications
| CD45.1 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Mouse | |
| 25 μg | |
| Cell Biology | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| A20 | |
| APC-R700 | |
| Ly-5; Lyt4; CD45R; LCA; Ptprc; Protein tyrosine phosphatase receptor type C | |
| SJL mouse thymocytes and splenocytes | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction