missing translation for 'onlineSavingsMsg'
Learn More

CD44 Std./H-CAM Ab-4 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunoprecipitation and flow cytometry experiments.

Brand:  Epredia MS-668-P1

 View more versions of this product

Product Code. 12691977

  • ONLINE EXCLUSIVE PRICE
    £322.00
    £301.89 / 500µL
    Save £20.11
    6% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD44 is a cell surface glycoprotein expressed on lymphocytes, monocytes, and granulocytes. CD44 is also known as homing cell adhesion molecule (H-CAM), Phagocytic glycoprotein-1 (PgP-1), ECM-III, HUTCH-1, or Hermes-1. CD44 has been implicated in cell migration, in lymphocyte homing, and in tumor metastasis. Ab-4 reacts with red cells pretreated with 2-aminoethylisothiouronium bromide (AET). Digestion of intact erythrocytes by trypsin treatment leaves a 50-54 kDa protein fragment attached to the membrane, while chymotrypsin leaves a somewhat similarly sized 51-56 kDa component that are recognized by Ab-4. This antibody enhances IL-2 production by CD2-transfected murine hybridoma stimulated by JY cells while it has no significant effect on IL-2 production by Jurkat cells stimulated with CD3, PMA, or CD2. Ab-4 enhances proliferation of PBL stimulated with CD2.

Host Species: Mouse

Clone: 156-3C11

Isotype: IgG2a

Species Reactivity: Human, Baboon and Green Monkey. Does not react with rat. Others not tested.

Epitope: Resistant to digestion by trypsin and chymotrypsin

Immunogen: Stimulated human leukocytes

Molecular Weight: 80-95 kDa

Positive Control: HUVEC or HeLa cells, tonsil or esophageal CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD44 Std./H-CAM Ab-4
Monoclonal
200μg/mL
Mouse
500 μL
Oncogenesis
Human, Non-human Primate
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
156-3C11
Unconjugated
Stimulated human leukocytes
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.