missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Rabbit anti-Human, Clone: 2V9T2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £395.00
Specifications
| Antigen | CD44 |
|---|---|
| Clone | 2V9T2 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18330413
|
Novus Biologicals
NBP3-15617-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18332127
|
Novus Biologicals
NBP3-15617-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD44 Monoclonal antibody specifically detects CD44 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDownSpecifications
| CD44 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human CD44 (P16070). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 2V9T2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 960 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title