missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (VFF-17), PerCP - variant 7/8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65535PCP
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD44 | |
| Monoclonal | |
| PerCP | |
| P16070 | |
| CD44 | |
| Made to Human CD44v7/8 | |
| 0.1 mL | |
| Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 960 | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| VFF-17 | |
| Flow Cytometry | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Specifically recognizes an epitope encoded by exons v7-v8 on the variant portion of human CD44. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction