missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (OX-50) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 11 publications
Brand: Novus Biologicals NB600-1317
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| CD44 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot reported in scientific literature (PMID 8537452), Flow Cytometry 1:50 - 1:100, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 31229690), Immunohistochemistry-Paraffin 1:50 - 1:100. Use reported in scientific literature (PMID 16673425), Immunohistochemistry-Frozen 1:50 - 1:100. Use reported in scientific literature (PMID 16673425), CyTOF-ready | |
| CD44 | |
| Rat T cell blasts from a mixed lymphocyte reaction between purified CD4+ T cells and irradiated spleen cells. | |
| 0.1 mL | |
| Cancer, Cell Biology, Extracellular Matrix, Hematopoietic Stem Cell Markers, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers | |
| 960 | |
| Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| OX-50 | |
| Unconjugated | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognizes the rat CD44 cell surface antigen, expressed by T cells, B cells, macrophages and thymocytes. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction