missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Antibody (CL13319) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18578IR
This item is not returnable.
View return policy
Description
CD44 Monoclonal antibody specifically detects CD44 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| CD44 | |
| Monoclonal | |
| DyLight 755 | |
| CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| This antibody was developed using a synthetic peptide derived from P16070, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Cancer, Cell Biology, Core ESC Like Genes, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 960 | |
| Store at 4⪚C in the dark. | |
| IgG3 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL13319 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction