missing translation for 'onlineSavingsMsg'
Learn More

CD437, MedChemExpress

Product Code. 16494366
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494366 100 mg 100mg
16452606 25 mg 25mg
16429636 50 mg 50mg
16403078 10 mg 10mg
16455807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494366 Supplier Medchem Express Supplier No. HY100532
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CD437 is a selective Retinoic Acid Receptor γ (RARγ) agonist.

CD437 is a selective Retinoic Acid Receptor γ (RARγ) agonist.

Specifications

Chemical Name or Material CD437
CAS 125316-60-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H26O3
Quantity 100 mg
Synonym AHPN
Solubility Information DMSO : 150 mg/mL (376.42 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC=C2C=C(C3=CC=C(O)C(C4(C5)CC6CC5CC(C6)C4)=C3)C=CC2=C1)O
Molecular Weight (g/mol) 398.49
Formula Weight 398.49
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.