missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43/Sialophorin Rabbit anti-Human, Clone: 84-3C1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-12075-0.2MG
This item is not returnable.
View return policy
Description
CD43/Sialophorin Monoclonal antibody specifically detects CD43/Sialophorin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| CD43/Sialophorin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43) | |
| This antibody was raised by immunizng mice with stimulated human leukocytes. | |
| 0.2 mg | |
| B Cell Development and Differentiation Markers, Immunology | |
| 6693 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| 84-3C1 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction