missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD43/Sialophorin Monoclonal antibody specifically detects CD43/Sialophorin in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD43/Sialophorin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 84-3C1 |
| Conjugate | PE |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43) |
| Host Species | Rabbit |
| Immunogen | This antibody was raised by immunizng mice with stimulated human leukocytes. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?