Learn More
CD43 Rat anti-Mouse, FITC, Clone: S7, BD
Rat Monoclonal Antibody
£186.68 - £431.58
Specifications
| Antigen | CD43 |
|---|---|
| Clone | S7 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15811409
|
BD Biosciences
561856 |
0.1 mg |
£194.00 £186.68 / Each Save £7.32 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15817118
|
BD Biosciences
553270 |
0.5 mg |
£449.00 £431.58 / Each Save £17.42 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The S7 monoclonal antibody specifically binds to the 115 kDa glycosylated form of CD43 (Ly-48, Leukosialin). CD43 is expressed on IL-7-responsive pro-B cells, plasma cells, peritoneal and splenic CD5+ B cells (B-1 cells), granulocytes, monocytes, macrophages, platelets, natural killer cells, thymocytes, peripheral T cytotoxic/suppressor cells, and most T helper cells, but not resting conventional peripheral B cells. CD43 expression has also been detected on pluripotent hematopoietic stem cells and myeloid, lymphoid, and NK-cell progenitors in the bone marrow. Studies of CD43-deficient mice indicate that CD43 participates in the negative regulation of T-cell activation and adhesion.
Specifications
| CD43 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Mouse Plasmacytoma MOPC-315 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| S7 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG2a κ | |
| Affinity Purified | |
| Spn ; Sialophorin; Leukosialin; Ly-48; Ly48; Galgp; LEUK |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.