Learn More
Invitrogen™ CD42b Recombinant Rabbit Monoclonal Antibody (6Y2I1)
Description
Immunogen sequence: HPHPICEVSK VASHLEVNCD KRNLTALPPD LPKDTTILHL SENLLYTFSL ATLMPYTRLT QLNLDRCELT KLQVDGTLPV LGTLDLSHNQ LQSLPLLGQT LPALTVLDVS FNRLTSLPLG ALRGLGELQE LYLKGNELKT LPPGLLTPTP KLEKLSLANN NLTELPAGLL NGLENLDTLL LQENSLYTIP KGFFGSHLLP FAFLHGNPWL CNCEILYFRR WLQDNAENVY VWKQGVDVKA MTSNVASVQC DNSDKFPVYK YPGKGCPTLG DEGDTDLYDY YPEEDTEGDK VRATRTVVKF PTKAHTTPWG LFYSWSTASL DSQMPSSLHP TQESTKEQTT FPPRWTPNFT LHMESITFSK TPKSTTEPTP SPTTSEPVPE PAPNMTTLEP TPSPTTPEPT SEPAPSPTTP EPTSEPAPSP TTPEPTSEPA PSPTTPEPTP IPTIATSPTI LVSATSLITP KSTFLTTTKP VSLLESTKKT IPELDQPPKL RGVLQGHLES SRNDPFLHPD FCCLLPL.
Specifications
Specifications
| Antigen | CD42b |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Recombinant Monoclonal |
| Clone | 6Y2I1 |
| Concentration | 1.01 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Gp1ba |
| Gene Accession No. | P07359 |
| Gene Alias | Antigen CD42b-alpha; BDPLT1; BDPLT3; BSS; CD42a; CD42B; CD42b-alpha; DBPLT3; Glycocalicin; glycoprotein 1b, alpha polypeptide; glycoprotein Ib (platelet), alpha polypeptide; glycoprotein Ib platelet alpha subunit; glycoprotein Ib platelet subunit alpha; glycoprotein Ibalpha; GP1B; GP1BA; GPIB; GPIb alpha; GP-Ib alpha; GPIbA; GPIbalpha; GPIX; GP-IX; MGC34595; mutant platelet membrane glycoprotein Ib-alpha; platelet glycoprotein Ib alpha chain; platelet glycoprotein Ib alpha polypeptide; Platelet GPIX; platelet membrane glycoprotein 1b-alpha subunit; platelet membrane glycoprotein Ib-alpha; VWDP |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.