missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD42b/GPIb alpha Antibody (HIP1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-511PE
This item is not returnable.
View return policy
Description
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human, Mouse, Primate samples. It is validated for Flow Cytometry.
Specifications
| CD42b/GPIb alpha | |
| Monoclonal | |
| PE | |
| Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody HIP1 reacts with CD42b (GPIb alpha), a 135-145 kDa membrane glycoprotein expressed on platelets and megakaryocytes. CD42b and CD42c (GPIb beta) are composed in a disulfide linked heterodimer (CD42b/c; 160 kDa); CD42b/c forms a noncovalent complex with CD42a and CD42d. HLDA IV; WS Code P 40 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| HIP1 | |
| Flow Cytometry | |
| GP1BA | |
| Peripheral blood mononuclear cells of a patient suffering with CLL. | |
| 0.1 mL | |
| Immunology | |
| 2811 | |
| Human, Mouse, Primate | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction