missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD42b/GPIb alpha Monoclonal antibody specifically detects CD42b/GPIb alpha in Human, Mouse, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
Specifications
Specifications
| Antigen | CD42b/GPIb alpha |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF |
| Classification | Monoclonal |
| Clone | HIP1 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit |
| Host Species | Mouse |
| Immunogen | Peripheral blood mononuclear cells of a patient suffering with CLL. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?