Learn More
CD42a Mouse anti-Human, PE, Clone: ALMA.16, BD
Mouse Monoclonal Antibody
£132.62 - £268.36
Specifications
| Antigen | CD42a |
|---|---|
| Clone | ALMA.16 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15881399
|
BD Biosciences
561853 |
25 Tests |
£139.00 £132.62 / Each Save £6.38 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15899628
|
BD Biosciences
558819 |
100 Tests |
£278.00 £268.36 / Each Save £9.64 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reacts with a 17-22kD, single chain integral membrane glycoprotein, also known as GPIX. CD42a forms a noncovalently linked complex (GPIb/GPIX/GPV) with CD42b, CD42c and CD42d. It is expressed on platelets and megakaryocytes and is absent on the platelets of patients with Bernadr-Soulier Syndrome (BSS). Although the CD42a function is not fully understood, GPIX glycoprotein is important for the assembly and membrane expression of the complex and for the maintenance of the functional conformation of CD42b (GPIb).
Specifications
| CD42a | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| ALMA.16 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.