missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD41a Monoclonal Antibody (HIP8), NovaFluor™ Yellow 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

£154.00 - £340.00

Specifications

Antigen CD41a
Clone HIP8
Concentration 0.6 μg/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30055841
View Documents
Invitrogen™
H059T03Y04-A
100 Tests
£340.00
Pack of 100
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30055294
View Documents
Invitrogen™
H059T02Y04-A
25 Tests
£154.00
Pack of 25
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD41a Monoclonal antibody specifically detects CD41a in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD41a
0.6 μg/Test
Monoclonal
Mouse
Human
P08514
3674
Primary
4°C, store in dark, DO NOT FREEZE!
HIP8
Flow Cytometry
NovaFluor Yellow 660
RUO
PBS with 0.09% sodium azide; pH 7.2
AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93
IgG1 κ
Affinity chromatography
Itga2b
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.