missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD41a Monoclonal Antibody (HIP8), eFluor™ 450, eBioscience™
GREENER_CHOICE

Mouse Monoclonal Antibody

£282.00

Specifications

Antigen CD41a
Clone HIP8
Concentration 5 μL/Test
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15549676
GREENER_CHOICE
View Documents
Invitrogen™
48-0419-42
100 Tests
£282.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The HIP8 monoclonal antibody reacts with the human CD41 molecule, the integrin alpha IIb also known as platelet GPIIb. CD41 non-covalently associates with integrin beta3 (GPIIIa, CD61) and is expressed by megakaryocytes and platelets. The CD41/CD61 complex is a receptor for fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin and mediates platelets aggregation. HIP8 blocks platelet aggregation. Applications Reported: This HIP8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HIP8 antibody has been pre-titrated and tested by flow cytometric analysis of human platelets. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD41, also known as platelet glycoprotein IIb or ITGA2B, is a protein composed of two subunits: a 120 kDa alpha subunit and a 23 kDa beta subunit. It interacts with CD61 (gpIIIa, integrin beta III) in the presence of calcium to form a functional adhesive protein receptor. Initially thought to be expressed exclusively on platelets and megakaryocytes, CD41 is also found on hematopoietic progenitors in the embryo, fetus, and adult, indicating its role in early stages of hematopoietic differentiation. CD41 plays a crucial role in platelet function and blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to various adhesion molecules, including von Willebrand factor, fibrinogen, fibronectin, and vitronectin, facilitating platelet adhesion and aggregation. This receptor is essential for platelet function, contributing to the binding of several adhesion molecules and playing a significant role in hemostasis. Diseases associated with CD41 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 bleeding disorder, highlighting its importance in normal platelet function and blood coagulation processes.
Specifications

Specifications

CD41a
5 μL/Test
Flow Cytometry
eFluor 450
Mouse
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
P08514
3674
Primary
Affinity chromatography
HIP8
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG1 κ
Human
Itga2b
AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93
Itga2b
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.