Learn More
CD41/Integrin alpha 2b Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24552-1-AP-150UL
 View more versions of this product
Product Code. 16893025
  • £349.00 / 150µL
Description
CD41 (platelet glycoprotein IIb, ITGA2B) is composed of two subunits -120 kDa a, alpha and 23 kDa b, beta- that interact with CD61 in the presence of calcium to form a functional adhesive protein receptor. CD41 is also involved in blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to a variety of proteins including von Willebrand factor, fibrinogen, fibronectin and vitronectin. CD41 is mainly expressed on megakaryocyte-platelet lineage, but generally belongs to the antigens that are expressed during the early stages of hematopoietic differentiation. Diseases associated with CD41 dysfunction include Glansmann Thrombasthenia, and Platelet type-16 bleeding disorder.
Specifications
Specifications
CD41/Integrin alpha 2b
Polyclonal
Unconjugated
ITGA2B
CD41, CD41B, GP2B, GPalpha IIb, GPIIb, GTA, HPA3, Integrin alpha IIb, ITGA2B, ITGAB
Rabbit
Antigen Affinity Chromatography
RUO
16399, 3674, 685269
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.45 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P08514, Q9QUM0
Itga2b
CD41/Integrin alpha 2b Fusion Protein Ag19459
150 μL
Primary
Human, Mouse, Rat