missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD41/CD61 Antibody (PAC-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62201
This item is not returnable.
View return policy
Description
CD41/CD61 Monoclonal antibody specifically detects CD41/CD61 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Flow (Cell Surface), Immunocytochemistry
Specifications
| CD41/CD61 | |
| Monoclonal | |
| Unconjugated | |
| TBS (pH 8.0) | |
| Mouse | |
| Multi-step | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Flow Cytometry (Cell Surface), Immunocytochemistry | |
| PAC-1 | |
| Flow Cytometry 2-4 μg/ml, Immunoprecipitation 1:10-1:500, Flow (Cell Surface), Immunocytochemistry | |
| CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
| Human platelets | |
| 0.1 mg | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
| 3674 | |
| Store at 4°C. Do not freeze. | |
| IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction