missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD41/CD61 Antibody (CAPP2A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-60899
This item is not returnable.
View return policy
Description
CD41/CD61 Monoclonal specifically detects CD41/CD61 in Bovine, Goat, Water Buffalo, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD41/CD61 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
| Mouse | |
| Unpurified | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
| 3674 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Functional Assay | |
| CAPP2A | |
| Unconjugated | |
| Ascites with 0.09% Sodium Azide | |
| ITGA2B | |
| Bovine platelets | |
| 0.1 mg | |
| Primary | |
| Bovine, Goat, Water Buffalo, Sheep | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction