missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40L/CD154 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16668-1-AP-20UL
This item is not returnable.
View return policy
Description
The CD40 antigen is a single chain glycoprotein. It is known to be a member of the tumor necrosis factor / nerve growth factor superfamily and shows a significant homology to the Hodgkin′s disease associated antigen, CD30. CD40 is present on all B cells except plasma cells. It is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. The precise function of the CD40 antigen is unknown but it appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.Specifications
| CD40L/CD154 | |
| Polyclonal | |
| Unconjugated | |
| CD40LG | |
| CD154, CD40 L, CD40 ligand, CD40L, CD40LG, gp39, hCD40L, HIGM1, IGM, IMD3, T BAM, T cell antigen Gp39, TNF related activation protein, TNFSF5, TRAP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21947, 959 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27548, P29965 | |
| CD40LG | |
| CD40L/CD154 Fusion Protein Ag10147 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction