missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD40/TNFRSF5 Antibody (sotigalimab) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | CD40/TNFRSF5 |
|---|---|
| Clone | sotigalimab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30045755
|
Novus Biologicals™
NBP3-28642-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048111
|
Novus Biologicals™
NBP3-28642-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047445
|
Novus Biologicals™
NBP3-28642-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| CD40/TNFRSF5 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| TNFRSF5/ CD40 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| sotigalimab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P25942 | |
| 958 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title