missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ CD40/TNFRSF5 Antibody (selicrelumab) - IgG2SA, Novus Biologicals™

Human Monoclonal Antibody

£162.00 - £695.00

Specifications

Antigen CD40/TNFRSF5
Clone selicrelumab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30045195
View Documents
Novus Biologicals™
NBP3-28787-100ug
100 μg
£247.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044296
View Documents
Novus Biologicals™
NBP3-28787-1mg
1 mg
£695.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045421
View Documents
Novus Biologicals™
NBP3-28787-50ug
50 μg
£162.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

CD40/TNFRSF5
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
TNFRSF5/ CD40
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
selicrelumab
Monoclonal
Purified
RUO
Human
P25942
958
IgG2
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.